missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Peptide deformylase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Peptide deformylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6476-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417745.1 | |
Peptide deformylase Protein | |
Research Use Only | |
def | |
Recombinant Protein | |
Yeast | |
SVLQVLHIPD ERLRKVAKPV EEVNAEIQRI VDDMFETMYA EEGIGLAATQ VDIHQRIIVI DVSENRDERL VLINPELLEK SGETGIEEGC LSIPEQRALV PRAEKVKIRA LDRDGKPFEL EADGLLAICI QHEMDHLVGK LFMDYLSPLK QQRIRQKVEK LDRLKARA | |
Greater than 90% as determined by SDS-PAGE. |
947780 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |