missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Peptide deformylase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Peptide deformylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6476-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083580

  • 2325.00 EUR / 500µg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417745.1
Peptide deformylase Protein
Research Use Only
def
Recombinant Protein
Yeast
SVLQVLHIPD ERLRKVAKPV EEVNAEIQRI VDDMFETMYA EEGIGLAATQ VDIHQRIIVI DVSENRDERL VLINPELLEK SGETGIEEGC LSIPEQRALV PRAEKVKIRA LDRDGKPFEL EADGLLAICI QHEMDHLVGK LFMDYLSPLK QQRIRQKVEK LDRLKARA
Greater than 90% as determined by SDS-PAGE.
947780
500 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers