missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Penicillin-binding protein 1A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Penicillin-binding protein 1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7405-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417855.4 | |
Penicillin-binding protein 1A Protein | |
Research Use Only | |
mrcA | |
Recombinant Protein | |
E. coli | |
MLDEGYITQQ QFDQTRTEAI NANYHAPEIA FSAPYLSEMV RQEMYNRYGE SAYEDGYRIY TTITRKVQQA AQQAVRNNVL DYDMRHGYRG PANVLWKVGE SAWDNNKITD TLKALPTYGP LLPAAVTSAN PQQATAMLAD GSTVALSMEG VRWARPYRSD TQQGPTPRKV TDVLQTGQQI WVRQVGDAWW LAQVPEVNSA LVSINPQNGA VMALVGGFDF NQSKFNRATQ ALRQVGSNIK PFLYTAAMDK GLTLASMLND VPISRWDASA GSDWQPKNSP PQYAGPIRLR QGLGQSKNVV M | |
Greater than 80% as determined by SDS-PAGE. |
947907 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |