missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Oxygen-insensitive NADPH nitroreductase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.
352.00 EUR - 3575.00 EUR
Specifications
Accession Number | NP_415372.1 |
---|---|
Gene ID (Entrez) | 945483 |
Name | Oxygen-insensitive NADPH nitroreductase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057350
|
enQuireBio™
QP7087-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077350
|
enQuireBio™
QP7087-YE-1MG |
1 mg |
3575.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067350
|
enQuireBio™
QP7087-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087350
|
enQuireBio™
QP7087-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097350
|
enQuireBio™
QP7087-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007360
|
enQuireBio™
QP7087-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415372.1 | |
Oxygen-insensitive NADPH nitroreductase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945483 | |
Research Use Only | |
nfsA | |
Recombinant Protein | |
Yeast | |
MTPTIELICG HRSIRHFTDE PISEAQREAI INSARATSSS SFLQCSSIIR ITDKALREEL VTLTGGQKHV AQAAEFWVFC ADFNRHLQIC PDAQLGLAEQ LLLGVVDTAM MAQNALIAAE SLGLGGVYIG GLRNNIEAVT KLLKLPQHVL PLFGLCLGWP ADNPDLKPRL PASILVHENS YQPLDKGALA QYDEQLAEYY LTRGSNNRRD TWSDHIRRTI IKESRPFILD YLHKQGWATR | |
Greater than 90% as determined by SDS-PAGE. |