missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ornithine carbamoyltransferase chain F Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ornithine carbamoyltransferase chain F, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_414807.1 |
---|---|
Gene ID (Entrez) | 944844 |
Name | Ornithine carbamoyltransferase chain F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006271
|
enQuireBio™
QP8748-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026271
|
enQuireBio™
QP8748-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016271
|
enQuireBio™
QP8748-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036271
|
enQuireBio™
QP8748-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046271
|
enQuireBio™
QP8748-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056271
|
enQuireBio™
QP8748-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_414807.1 | |
Ornithine carbamoyltransferase chain F Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
944844 | |
Research Use Only | |
argF | |
Recombinant Protein | |
E. coli | |
SDLYKKHFLK LLDFTPAQFT SLLTLAAQLK ADKKNGKEVQ KLTGKNIALI FEKDSTRTRC SFEVAAFDQG ARVTYLGPSG SQIGHKESIK DTARVLGRMY DGIQYRGHGQ EVVETLAQYA GVPVWNGLTN EFHPTQLLAD LMTMQEHLPG KAFNEMTLVY AGDARNNMGN SMLEAAALTG LDLRLLAPKA CWPEESLVAE CSALAEKHGG KITLTEDVAA GVKGADFIYT DVWVSMGEAK EKWAERIALL RGYQVNAQMM ALTDNPNVKF LHCLPAFHDD QTTLGKQMAK EFDLHGGMEV TDEVFESAAS IVFDQAENRM HTIKAVMMAT LGE | |
Greater than 90% as determined by SDS-PAGE. |