missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli nrdB Protein
A DNA sequence encoding the Escherichia coli (strain K12) nrdB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6953-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_416738.1 | |
nrdB Protein | |
Research Use Only | |
nrdB | |
Recombinant Protein | |
Yeast | |
AYTTFSQTKN DQLKEPMFFG QPVNVARYDQ QKYDIFEKLI EKQLSFFWRP EEVDVSRDRI DYQALPEHEK HIFISNLKYQ TLLDSIQGRS PNVALLPLIS IPELETWVET WAFSETIHSR SYTHIIRNIV NDPSVVFDDI VTNEQIQKRA EGISSYYDEL IEMTSYWHLL GEGTHTVNGK TVTVSLRELK KKLYLCLMSV NALEAIRFYV SFACSFAFAE RELMEGNAKI IRLIARDEAL HLTGTQHMLN LLRSGADDPE MAEIAEECKQ ECYDLFVQAA QQEKDWADYL FRDGSMIGLN KDILCQYVEY ITNIRMQAVG LDLPFQTRSN PIPWINTWLV SDNVQVAPQE VEVSSYLVGQ IDSEVDTDDL SNFQL | |
Greater than 90% as determined by SDS-PAGE. |
946732 | |
1 mg | |
Not determined. | |
Full Length | |
Yeast | |
His | |
Tris-based buffer, 50% glycerol |