missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli NAD(P)H-flavin reductase Protein

A DNA sequence encoding the Escherichia coli (strain K12) NAD(P)H-flavin reductase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8942-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047261

  • 352.00 EUR / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418286.1
NAD(P)H-flavin reductase Protein
Research Use Only
fre
Recombinant Protein
E. coli
TTLSCKVTSV EAITDTVYRV RIVPDAAFSF RAGQYLMVVM DERDKRPFSM ASTPDEKGFI ELHIGASEIN LYAKAVMDRI LKDHQIVVDI PHGEAWLRDD EERPMILIAG GTGFSYARSI LLTALARNPN RDITIYWGGR EEQHLYDLCE LEALSLKHPG LQVVPVVEQP EAGWRGRTGT VLTAVLQDHG TLAEHDIYIA GRFEMAKIAR DLFCSERNAR EDRLFGDAFA FI
Greater than 90% as determined by SDS-PAGE.
948325
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers