missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Murein tetrapeptide carboxypeptidase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Murein tetrapeptide carboxypeptidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6956-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006890

  • 1315.00 EUR / 500µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_415710.1
Murein tetrapeptide carboxypeptidase Protein
Research Use Only
ldcA
Recombinant Protein
E. coli
MSLFHLIAPS GYCIKQHAAL RGIQRLTDAG HQVNNVEVIA RRCERFAGTE TERLEDLNSL ARLTTPNTIV LAVRGGYGAS RLLADIDWQA LVARQQHDPL LICGHSDFTA IQCGLLAHGN VITFSGPMLV ANFGADELNA FTEHHFWLAL RNETFTIEWQ GEGPTCRAEG TLWGGNLAML ISLIGTPWMP KIENGILVLE DINEHPFRVE RMLLQLYHAG ILPRQKAIIL GSFSGSTPND YDAGYNLESV YAFLRSRLSI PLITGLDFGH EQRTVTLPLG AHAILNNTRE GTQLTISGHP VLKM
Greater than 90% as determined by SDS-PAGE.
945756
500 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers