missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Modification methylase EcoRV Protein

A DNA sequence encoding the Escherichia coli (strain K12) Modification methylase EcoRV, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7415-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048760

  • 865.00 EUR / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_863581.1
Modification methylase EcoRV Protein
Research Use Only
ecoRVM
Recombinant Protein
E. coli
MKDKVFVPPI KSQGIKTKLV PCIKRIVPKN FNGVWVEPFM GTGVVAFNVA PKDALLCDTN PHLISFYNAL KNKDITGDLV KDFLYREGEK LLLSNGEYYY EVRERFNNYK EPLDFLFLNR SCFNGMIRFN SKGGFNVPFC KKPNRFAQAY ITKISNQVDR ISEIISKGNY TFLCQSFEKT IGMVNRDDVV YCDPPYIGRH VDYFNSWGER DERLLFETLS SLNATFITST WHHNDYRENK YVRDLWSSFR ILTKEHFYHV GASEKNRSPM VEALITNIAK DIIDHIEKSS GDILVIEE
Greater than 90% as determined by SDS-PAGE.
1446622
100 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers