missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Lactose operon repressor Protein
A DNA sequence encoding the Escherichia coli (strain K12) Lactose operon repressor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7406-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_414879.3 | |
Lactose operon repressor Protein | |
Research Use Only | |
lacI | |
Recombinant Protein | |
E. coli | |
MKPVTLYDVA EYAGVSYQTV SRVVNQASHV SAKTREKVEA AMAELNYIPN RVAQQLAGKQ SLLIGVATSS LALHAPSQIV AAIKSRADQL GASVVVSMVE RSGVEACKAA VHNLLAQRVS GLIINYPLDD QDAIAVEAAC TNVPALFLDV SDQTPINSII FSHEDGTRLG VEHLVALGHQ QIALLAGPLS SVSARLRLAG WHKYLTRNQI QPIAEREGDW SAMSGFQQTM QMLNEGIVPT AMLVANDQMA LGAMRAITES GLRVGADISV VGYDDTEDSS CYIPPLTTIK QDFRLLGQTS VDRLLQLSQG QAVKGNQLLP VSLVKRKTTL APNTQTASPR ALADSLMQLA RQVSRLESGQ | |
Greater than 90% as determined by SDS-PAGE. |
945007 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |