missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli GTP cyclohydrolase-2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) GTP cyclohydrolase-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8879-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415793.1 | |
GTP cyclohydrolase-2 Protein | |
Research Use Only | |
ribA | |
Recombinant Protein | |
E. coli | |
MQLKRVAEAK LPTPWGDFLM VGFEELATGH DHVALVYGDI SGHTPVLARV HSECLTGDAL FSLRCDCGFQ LEAALTQIAE EGRGILLYHR QEGRNIGLLN KIRAYALQDQ GYDTVEANHQ LGFAADERDF TLCADMFKLL GVNEVRLLTN NPKKVEILTE AGINIVERVP LIVGRNPNNE HYLDTKAEKM GHLLNK | |
Greater than 90% as determined by SDS-PAGE. |
945763 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |