missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glutaminase 1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutaminase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6951-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415018.1 | |
Glutaminase 1 Protein | |
Research Use Only | |
glsA1 | |
Recombinant Protein | |
E. coli | |
MLDANKLQQA VDQAYTQFHS LNGGQNADYI PFLANVPGQL AAVAIVTCDG NVYSAGDSDY RFALESISKV CTLALALEDV GPQAVQDKIG ADPTGLPFNS VIALELHGGK PLSPLVNAGA IATTSLINAE NVEQRWQRIL HIQQQLAGEQ VALSDEVNQS EQTTNFHNRA IAWLLYSAGY LYCDAMEACD VYTRQCSTLL NTIELATLGA TLAAGGVNPL THKRVLQADN VPYILAEMMM EGLYGRSGDW AYRVGLPGKS GVGGGILAVV PGVMGIAAFS PPLDEDGNSV RGQKMVASVA KQLGYNVFKG | |
Greater than 90% as determined by SDS-PAGE. |
946187 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-GST | |
Tris-based buffer, 50% glycerol |