missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli fkpA Protein

A DNA sequence encoding the Escherichia coli (strain K12) fkpA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7261-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087920

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000838261.1
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
fkpA Protein
Research Use Only
fkpA
Recombinant Protein
Yeast
AEAAKPATTA DSKAAFKNDD QKSAYALGAS LGRYMENSLK EQEKLGIKLD KDQLIAGVQD AFADKSKLSD QEIEQTLQAF EARVKSSAQA KMEKDAADNE AKGKEYREKF AKEKGVKTSS TGLVYQVVEA GKGEAPKDSD TVVVNYKGTL IDGKEFDNSY TRGEPLSFRL DGVIPGWTEG LKNIKKGGKI KLVIPPELAY GKAGVPGIPP NSTLVFDVEL LDVKPAPKAD AKPEADAKAA DSAKK
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers