missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cystathionine beta-lyase MetC Protein

A DNA sequence encoding the Escherichia coli (strain K12) Cystathionine beta-lyase MetC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8801-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006541

  • 865.00 EUR / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417481.1
Cystathionine beta-lyase MetC Protein
Research Use Only
metC
Recombinant Protein
E. coli
MADKKLDTQL VNAGRSKKYT LGAVNSVIQR ASSLVFDSVE AKKHATRNRA NGELFYGRRG TLTHFSLQQA MCELEGGAGC VLFPCGAAAV ANSILAFIEQ GDHVLMTNTA YEPSQDFCSK ILSKLGVTTS WFDPLIGADI VKHLQPNTKI VFLESPGSIT MEVHDVPAIV AAVRSVVPDA IIMIDNTWAA GVLFKALDFG IDVSIQAATK YLVGHSDAMI GTAVCNARCW EQLRENAYLM GQMVDADTAY ITSRGLRTLG VRLRQHHESS LKVAEWLAEH PQVARVNHPA LPGSKGHEFW KRDFTGSSGL FSFVLKKKLN NEELANYLDN FSLFSMAYSW GGYESLILAN QPEHIAAIRP QGEIDFSGTL IRLHIGLEDV DDLIADLDAG FA
Greater than 90% as determined by SDS-PAGE.
946240
100 μg
Not determined.
Partial (see sequence information for more details).
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers