missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cystathionine beta-lyase MetC Protein
A DNA sequence encoding the Escherichia coli (strain K12) Cystathionine beta-lyase MetC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8801-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417481.1 | |
Cystathionine beta-lyase MetC Protein | |
Research Use Only | |
metC | |
Recombinant Protein | |
E. coli | |
MADKKLDTQL VNAGRSKKYT LGAVNSVIQR ASSLVFDSVE AKKHATRNRA NGELFYGRRG TLTHFSLQQA MCELEGGAGC VLFPCGAAAV ANSILAFIEQ GDHVLMTNTA YEPSQDFCSK ILSKLGVTTS WFDPLIGADI VKHLQPNTKI VFLESPGSIT MEVHDVPAIV AAVRSVVPDA IIMIDNTWAA GVLFKALDFG IDVSIQAATK YLVGHSDAMI GTAVCNARCW EQLRENAYLM GQMVDADTAY ITSRGLRTLG VRLRQHHESS LKVAEWLAEH PQVARVNHPA LPGSKGHEFW KRDFTGSSGL FSFVLKKKLN NEELANYLDN FSLFSMAYSW GGYESLILAN QPEHIAAIRP QGEIDFSGTL IRLHIGLEDV DDLIADLDAG FA | |
Greater than 90% as determined by SDS-PAGE. |
946240 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |