missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Bifunctional protein HldE Protein

A DNA sequence encoding the Escherichia coli (strain K12) Bifunctional protein HldE, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6949-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026840

  • 2070.00 EUR / 500µg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417524.1
Bifunctional protein HldE Protein
500 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
947548
Greater than 90% as determined by SDS-PAGE.
Research Use Only
hldE
Recombinant Protein
E. coli
MKVTLPEFER AGVMVVGDVM LDRYWYGPTS RISPEAPVPV VKVNTIEERP GGAANVAMNI ASLGANARLV GLTGIDDAAR ALSKSLADVN VKCDFVSVPT HPTITKLRVL SRNQQLIRLD FEEGFEGVDP QPLHERINQA LSSIGALVLS DYAKGALASV QQMIQLARKA GVPVLIDPKG TDFERYRGAT LLTPNLSEFE AVVGKCKTEE EIVERGMKLI ADYELSALLV TRSEQGMSLL QPGKAPLHMP TQAQEVYDVT GAGDTVIGVL AATLAAGNSL EEACFFANAA AGVVVGKLGT STVSPIELEN AVRGRADTGF GVMTEEELKL AVAAARKRGE KVVMTNGVFD ILHAGHVSYL ANARKLGDRL IVAVNSDAST KRLKGDSRPV NPLEQRMIVL GALEAVDWVV SFEEDTPQRL IAGILPDLLV KGGDYKPEEI AGSKEVWANG GEVLVLNFED GCSTTNIIKK IQQDKKG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers