missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Beta-lactamase CTX-M-1 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase CTX-M-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7160-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007510

  • 760.00 EUR / 100µg


Estimated Shipment
21-02-2020
Add to basket

Specifications

Specifications

WP_013188473.1
Beta-lactamase CTX-M-1 Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
13909205
Greater than 90% as determined by SDS-PAGE.
Research Use Only
bla
Recombinant Protein
E. coli
QTADVQQKLA ELERQSGGRL GVALINTADN SQILYRADER FAMCSTSKVM AVAAVLKKSE SEPNLLNQRV EIKKSDLVNY NPIAEKHVDG TMSLAELSAA ALQYSDNVAM NKLISHVGGP ASVTAFARQL GDETFRLDRT EPTLNTAIPG DPRDTTSPRA MAQTLRNLTL GKALGDSQRA QLVTWMKGNT TGAASIQAGL PASWVVGDKT GSGDYGTTND IAVIWPKDRA PLILVTYFTQ PQPKAESRRD VLASAAKIVT NGL
Videos
Documents