missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecQ Protein
A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase RecQ, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8915-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000035581.1 | |
ATP-dependent DNA helicase RecQ Protein | |
Research Use Only | |
recQ | |
Recombinant Protein | |
E. coli | |
AQAEVLNLES GAKQVLQETF GYQQFRPGQE EIIDTVLSGR DCLVVMPTGG GKSLCYQIPA LLLNGLTVVV SPLISLMKDQ VDQLQANGVA AACLNSTQTR EQQLEVMTGC RTGQIRLLYI APERLMLDNF LEHLAHWNPV LLAVDEAHCI SQWGHDFRPE YAALGQLRQR FPTLPFMALT ATADDTTRQD IVRLLGLNDP LIQISSFDRP NIRYMLMEKF KPLDQLMRYV QEQRGKSGII YCNSRAKVED TAARLQSKGI SAAAYHAGLE NNVRADVQEK FQRDDLQIVV ATVAFGMGIN KPNVRFVVHF DIPRNIESYY QETGRAGRDG LPAEAMLFYD PADMAWLRRC LEEKPQGQLQ DIERHKLNAM GAFAEAQTCR RLVLLNYFGE GRQEPCGNCD ICLDPPKQYD GSTDAQIALS TIGRVNQRFG MGYVVEVIRG ANNQRIRDYG HDKLKVYGMG RDKSHEHWVS VIRQLIHLGL VTQNIAQHSA LQLTEAARPV LRGESSLQLA VPRIVALKPK AMQKSFGGNY DRKLFAKLRK LRKSIADESN VPPYVVFNDA TLIEMAEQMP ITASEMLSVN GVGMRKLERF GKPFMALIRA HVDGD | |
Greater than 90% as determined by SDS-PAGE. |
948318 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |