missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 4-hydroxy-tetrahydrodipicolinate synthase Protein

A DNA sequence encoding the Escherichia coli (strain K12) 4-hydroxy-tetrahydrodipicolinate synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8941-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087251

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_416973.1
4-hydroxy-tetrahydrodipicolinate synthase Protein
Research Use Only
dapA
Recombinant Protein
E. coli
MFTGSIVAIV TPMDEKGNVC RASLKKLIDY HVASGTSAIV SVGTTGESAT LNHDEHADVV MMTLDLADGR IPVIAGTGAN ATAEAISLTQ RFNDSGIVGC LTVTPYYNRP SQEGLYQHFK AIAEHTDLPQ ILYNVPSRTG CDLLPETVGR LAKVKNIIGI KEATGNLTRV NQIKELVSDD FVLLSGDDAS ALDFMQLGGH GVISVTANVA ARDMAQMCKL AAEGHFAEAR VINQRLMPLH NKLFVEPNPI PVKWACKELG LVATDTLRLP MTPITDSGRE TVRAALKHAG LL
Greater than 90% as determined by SDS-PAGE.
946952
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers