missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Epididymal secretory protein E1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Epididymal secretory protein E1, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Name | Epididymal secretory protein E1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NPC |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019900
|
enQuireBio™
QP7631-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039900
|
enQuireBio™
QP7631-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16029900
|
enQuireBio™
QP7631-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049900
|
enQuireBio™
QP7631-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059900
|
enQuireBio™
QP7631-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069900
|
enQuireBio™
QP7631-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Epididymal secretory protein E1 Protein | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NPC | |
Recombinant Protein | |
E. coli | |
VHFKDCGSAV GVIKELNVNP CPAQPCKLHK GQSYSVNVTF TSNIPSQSSK AVVHGIVLGV AVPFPIPEAD GCKSGINCPI QKDKTYSYLN KLPVKNEYPS IKLVVQWMLL GDNNQHLFCW EIPVQIEG | |
Greater than 90% as determined by SDS-PAGE. |