missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Caveolin-1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Caveolin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9031-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003296.1 | |
Caveolin-1 Protein | |
Research Use Only | |
CAV1 | |
Recombinant Protein | |
Yeast | |
MSGGKYVDSE GHLYTVPIRE QGNIYKPNNK AMAEEMSEKQ VYDAHTKEID LVNRDPKHLN DDVVKIDFED VIAEPEGTHS FDGIWKASFT TFTVTKYWFY RLLSALFGIP MALIWGIYFA ILSFLHIWAV VPCIKSFLIE IQCISRVYSI YVHTFCDPFF EAVGKIFSNI RINMQKET | |
Greater than 90% as determined by SDS-PAGE. |
403980 | |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |