missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7086-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Uncharacterized protein UL131 Protein | |
Research Use Only | |
UL131 | |
Recombinant Protein | |
E. coli | |
MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR | |
Greater than 80% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |