missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Uncharacterized protein UL131 Protein

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Uncharacterized protein UL131, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7086-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057340

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Uncharacterized protein UL131 Protein
Research Use Only
UL131
Recombinant Protein
E. coli
MCMMSHNKAF FLSLQHAAVS GVAVCLSVRR GAGSVPAGNR GKKTIITEYR ITGTRALARC PTKPVTSMWN SSWTSR
Greater than 80% as determined by SDS-PAGE.
200 μg
Not determined.
Full Length
Cytomegalovirus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers