missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster Peroxiredoxin 1 / PRDX1 Protein
A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Peroxiredoxin 1 / PRDX1, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001233694.1 |
---|---|
Gene ID (Entrez) | 100689332 |
Name | Peroxiredoxin 1 / PRDX1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022761
|
enQuireBio™
QP8117-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042761
|
enQuireBio™
QP8117-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032761
|
enQuireBio™
QP8117-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052761
|
enQuireBio™
QP8117-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062761
|
enQuireBio™
QP8117-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072761
|
enQuireBio™
QP8117-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001233694.1 | |
Peroxiredoxin 1 / PRDX1 Protein | |
Not determined. | |
Full Length | |
Chinese Hamster | |
His | |
Tris-based buffer, 50% glycerol |
100689332 | |
Research Use Only | |
PRDX1 | |
Recombinant Protein | |
E. coli | |
MSSGNAKIGY PAPNFKATAV MPDGQFRDIC LSEYRGKYVV FFFYPLDFTF VCPTEIIAFS DRAEEFKKLN CQVIGASVDS HFCHLAWINT PKKQGGLGPM NIPLVSDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITINDLPVG RSVDEILRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVQK SKEYFSKQK | |
Greater than 90% as determined by SDS-PAGE. |