missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster P4HB / ERBA2L Protein
A DNA sequence encoding the Chinese hamster P4HB / ERBA2L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7857-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001233622.1 | |
P4HB / ERBA2L Protein | |
Research Use Only | |
P4hb | |
Recombinant Protein | |
E. coli | |
DAPEEEDNVL VLKKSNFAEA LAAHNYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE TLIDSSEVAV IGFFKDVESD SAKQFLLAAE AVDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLGNFKK AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ATADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDVDLE EALEPDMEED DDQKAVKDEL | |
Greater than 90% as determined by SDS-PAGE. |
100689433 | |
500 μg | |
Not determined. | |
Full Length | |
Chinese Hamster | |
His | |
Tris-based buffer, 50% glycerol |