missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster C-C motif chemokine Protein
A DNA sequence encoding the Cricetulus griseus C-C motif chemokine, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8949-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
XP_003495840.1 | |
C-C motif chemokine Protein | |
Research Use Only | |
Full Length of Mature Protein | |
Chinese Hamster | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
100763833 | |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
QPDAVNSPLT CCYSFTAKRI PEKRLESYKR ITSSKCPKEA VIFITKLKRE ICADPKQDWV QTYTKKLDQS QAKSEAATVY KTAPLNANLT HESAVNASTT AFPTTDLRTS VRVTSMTVN | |
Greater than 90% as determined by SDS-PAGE. |