missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chimpanzee LTB / TNFSF3 / Lymphotoxin beta Protein

A DNA sequence encoding the Pan troglodytes (Chimpanzee) LTB / TNFSF3 / Lymphotoxin beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7788-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001070252.1
LTB / TNFSF3 / Lymphotoxin beta Protein
Research Use Only
LTB
Recombinant Protein
E. coli
QDQGGLVTET ADPGAQAQQG LGFQKLPEEE PETDLSPGLP AAHLIGAPLK GQGLGWETTK EQAFLTSGTQ FSDAEGLALP QDGLYYLYCL VGYRGRTPPG GGDPQGRSVT LRSSLYRAGG AYGPGTPELL LEGAETVTPV LDPARRQGYG PLWYTSVGFG GLVQLRRGER VYVNISHPDM VDFARGKTFF GAVMVG
Greater than 90% as determined by SDS-PAGE.
744486
10 μg
Not determined.
Extracellular Domain
Chimpanzee
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers