Learn More
enQuireBio™ Recombinant Chicken Vesicle-associated membrane protein 1
A DNA sequence encoding the Gallus gallus (Chicken) Vesicle-associated membrane protein 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6875-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001026292.1 | |
Vesicle-associated membrane protein 1 | |
Research Use Only | |
VAMP7 | |
Recombinant Protein | |
E. coli | |
AILFAVVARG TTILAKHAWC GGNFLEVTEQ ILAKIPSENN KLTYSHGNYL FHYICQDRII YLCITDDDFE RSRAFNFLNE IKKRFQTTYG SRAQTALPYA MNSEFSSVLA AQLKYHSESK GTDQVAETQA QVDELKGIMV RNIDLVAQRG EKLELLIDKT ENLVDSSVTF KTTSRNLARA | |
Greater than 90% as determined by SDS-PAGE. |
422297 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |