missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Riboflavin-binding Protein

A DNA sequence encoding the Gallus gallus (Chicken) Riboflavin-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8851-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006831

  • 437.00 EUR / 50µg


Estimated Shipment
13-02-2020
Add to basket

Specifications

Specifications

NP_990794.1
Riboflavin-binding Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Chicken
His
Tris-based buffer, 50% glycerol
396449
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RBP
Recombinant Protein
E. coli
QQYGCLEGDT HKANPSPEPN MHECTLYSES SCCYANFTEQ LAHSPIIKVS NSYWNRCGQL SKSCEDFTKK IECFYRCSPH AARWIDPRYT AAIQSVPLCQ SFCDDWYEAC KDDSICAHNW LTDWERDESG ENHCKSKCVP YSEMYANGTD MCQSMWGESF KVSESSCLCL QMNKKDMVAI KHLLSESSEE SSSMSSSEEH ACQKKLLK
Videos
Documents