missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Ovoinhibitor Protein
A DNA sequence encoding the Gallus gallus (Chicken) Ovoinhibitor, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Name | Ovoinhibitor Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | OIH |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047070
|
enQuireBio™
QP7020-YE-100UG |
100 μg |
965.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16087070
|
enQuireBio™
QP7020-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057070
|
enQuireBio™
QP7020-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077070
|
enQuireBio™
QP7020-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097070
|
enQuireBio™
QP7020-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067070
|
enQuireBio™
QP7020-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Ovoinhibitor Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
OIH | |
Recombinant Protein | |
Yeast | |
VNCSLYASGI GKDGTSWVAC PRNLKPVCGT DGSTYSNECG ICLYNREHGA NVEKEYDGEC RPKHVMIDCS PYLQVVRDGN TMVACPRILK PVCGSDSFTY DNECGICAYN AEHHTNISKL HDGECKLEIG SVDCSKYPST VSKDGRTLVA CPRILSPVCG TDGFTYDNEC GICAHNAEQR THVSKKHDGK CRQEIPEIDC DQYPTRKTTG GKLLVRCPRI LLPVCGTDGF TYDNECGICA HNAQHGTEVK KSHDGRCKER STPLDCTQYL SNTQNGEAIT ACPFILQEVC GTDGVTYSND CSLCAHNIEL GTSVAKKHDG RCREEVPELD CSKYKTSTLK DGRQVVACTM IYDPVCATNG VTYASECTLC AHNLEQRTNL GKRKNGRCEE DITKEHCREF QKVSPICTME YVPHCGSDGV TYSNRCFFCN AYVQSNRTLN LVSMAAC | |
Greater than 90% as determined by SDS-PAGE. |