missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Ovoinhibitor Protein
A DNA sequence encoding the Gallus gallus (Chicken) Ovoinhibitor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7020-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Ovoinhibitor Protein | |
Research Use Only | |
OIH | |
Recombinant Protein | |
E. coli | |
VNCSLYASGI GKDGTSWVAC PRNLKPVCGT DGSTYSNECG ICLYNREHGA NVEKEYDGEC RPKHVMIDCS PYLQVVRDGN TMVACPRILK PVCGSDSFTY DNECGICAYN AEHHTNISKL HDGECKLEIG SVDCSKYPST VSKDGRTLVA CPRILSPVCG TDGFTYDNEC GICAHNAEQR THVSKKHDGK CRQEIPEIDC DQYPTRKTTG GKLLVRCPRI LLPVCGTDGF TYDNECGICA HNAQHGTEVK KSHDGRCKER STPLDCTQYL SNTQNGEAIT ACPFILQEVC GTDGVTYSND CSLCAHNIEL GTSVAKKHDG RCREEVPELD CSKYKTSTLK DGRQVVACTM IYDPVCATNG VTYASECTLC AHNLEQRTNL GKRKNGRCEE DITKEHCREF QKVSPICTME YVPHCGSDGV TYSNRCFFCN AYVQSNRTLN LVSMAAC | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |