missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Anosmin-1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Anosmin-1, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_990755.1 |
---|---|
Gene ID (Entrez) | 396395 |
Name | Anosmin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031920
|
enQuireBio™
QP6250-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051920
|
enQuireBio™
QP6250-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041920
|
enQuireBio™
QP6250-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061920
|
enQuireBio™
QP6250-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071920
|
enQuireBio™
QP6250-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081920
|
enQuireBio™
QP6250-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_990755.1 | |
Anosmin-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |
396395 | |
Research Use Only | |
ANOS1 | |
Recombinant Protein | |
E. coli | |
SPAGPGAATA RRQDEAFSTA RCTSRCLSLQ ITRISAFFKH FQNNGSLAWC QNHKQCSKCL EPCKESWDLK KNHCQSFCEP LFPKKNYECL TSCEFLKYIL SVKQGDCPAP EKASGFAAAC VESCEADSEC SGVKKCCSNG CGHTCQVPKN LYKGVPLKPR KELKFIELQS GDLEVKWSSK FNISIEPVIY VVQRRWNQGI HPSEDDATNW QTVAQTTDER VQLSDIRASR WYQFRVAAVN VHGTRGFTAP SKHFRSSKDP | |
Greater than 90% as determined by SDS-PAGE. |