missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis Major outer membrane porin Protein

A DNA sequence encoding the Chlamydia trachomatis serovar L2 (strain 434/Bu / ATCC VR-902B) Major outer membrane porin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7292-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_009873292.1
Major outer membrane porin Protein
Research Use Only
ompA
Recombinant Protein
E. coli
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLQTDVNKEF QMGAKPTTAT GNAAAPSTCT ARENPAYGRH MQDAEMFTNA AYMALNIWDR FDVFCTLGAT SGYLKGNSAS FNLVGLFGDN ENHATVSDSK LVPNMSLDQS VVELYTDTTF AWSAGARAAL WECGCATLGA SFQYAQSKPK VEELNVLCNA AEFTINKPKG YVGQEFPLDL KAGTDGVTGT KDASIDYHEW QASLALSYRL NMFTPYIGVK WSRASFDADT IRIAQPKSAT TVFDVTTLNP TIAGAGDVKA SAEGQLGDTM QIVSLQLNKM KSRKSCGIAV GTTIVDADKY AVTVETRLID ERAAHVNAQF RF
Greater than 90% as determined by SDS-PAGE.
5858320
200 μg
Not determined.
Full Length
C. trachomatis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers