missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin Protein
A DNA sequence encoding the Chlamydia trachomatis serovar L2 (strain 434/Bu / ATCC VR-902B) Major outer membrane porin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7292-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_009873292.1 | |
Major outer membrane porin Protein | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLQTDVNKEF QMGAKPTTAT GNAAAPSTCT ARENPAYGRH MQDAEMFTNA AYMALNIWDR FDVFCTLGAT SGYLKGNSAS FNLVGLFGDN ENHATVSDSK LVPNMSLDQS VVELYTDTTF AWSAGARAAL WECGCATLGA SFQYAQSKPK VEELNVLCNA AEFTINKPKG YVGQEFPLDL KAGTDGVTGT KDASIDYHEW QASLALSYRL NMFTPYIGVK WSRASFDADT IRIAQPKSAT TVFDVTTLNP TIAGAGDVKA SAEGQLGDTM QIVSLQLNKM KSRKSCGIAV GTTIVDADKY AVTVETRLID ERAAHVNAQF RF | |
Greater than 90% as determined by SDS-PAGE. |
5858320 | |
50 μg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |