missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis 60 kDa chaperonin Protein
A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) 60 kDa chaperonin, was expressed in the hosts and tags indicated.
385.00 EUR - 3285.00 EUR
Specifications
Accession Number | NP_219613.1 |
---|---|
Gene ID (Entrez) | 884030 |
Name | 60 kDa chaperonin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077020
|
enQuireBio™
QP7010-10UG |
10 μg |
385.00 EUR
10µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017030
|
enQuireBio™
QP7010-50UG |
50 μg |
555.00 EUR
50µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067020
|
enQuireBio™
QP7010-100UG |
100 μg |
940.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097020
|
enQuireBio™
QP7010-200UG |
200 μg |
1550.00 EUR
200µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007030
|
enQuireBio™
QP7010-500UG |
500 μg |
2070.00 EUR
500µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087020
|
enQuireBio™
QP7010-1MG |
1 mg |
3285.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_219613.1 | |
60 kDa chaperonin Protein | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
884030 | |
Research Use Only | |
groL | |
Recombinant Protein | |
E. coli | |
MVAKNIKYNE EARKKIQKGV KTLAEAVKVT LGPKGRHVVI DKSFGSPQVT KDGVTVAKEV ELADKHENMG AQMVKEVASK TADKAGDGTT TATVLAEAIY TEGLRNVTAG ANPMDLKRGI DKAVKVVVDQ IRKISKPVQH HKEIAQVATI SANNDAEIGN LIAEAMEKVG KNGSITVEEA KGFETVLDIV EGMNFNRGYL SSYFATNPET QECVLEDALV LIYDKKISGI KDFLPVLQQV AESGRPLLII AEDIEGEALA TLVVNRIRGG FRVCAVKAPG FGDRRKAMLE DIAILTGGQL ISEELGMKLE NANLAMLGKA KKVIVSKEDT TIVEGMGEKE ALEARCESIK KQIEDSSSDY DKEKLQERLA KLSGGVAVIR VGAATEIEMK EKKDRVDDAQ HATIAAVEEG ILPGGGTALI RCIPTLEAFL PMLTNEDEQI GARIVLKALS APLKQIAANA GKEGAIIFQQ VMSRSANEGY DALRDAYTDM LEAGILDPAK VTRSALESAA SVAGLLLTTE ALIAEIPEEK PAAAPAMPGA GMDY | |
Greater than 90% as determined by SDS-PAGE. |