missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Flagellin Protein
A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) Flagellin, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | WP_011987021.1 |
---|---|
Gene ID (Entrez) | 5204320 |
Name | Flagellin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096740
|
enQuireBio™
QP6926-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016750
|
enQuireBio™
QP6926-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006750
|
enQuireBio™
QP6926-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026750
|
enQuireBio™
QP6926-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036750
|
enQuireBio™
QP6926-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046750
|
enQuireBio™
QP6926-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_011987021.1 | |
Flagellin Protein | |
Not determined. | |
Full Length | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5204320 | |
Research Use Only | |
CBO2730 | |
Recombinant Protein | |
E. coli | |
MIINHNLNAM NAHRQMAINT GNNGKAIEKL SSGLRINRAG DDAAGLAISE KMRGQIRGLN QASRNSQDAI SLIQTAEGAL NETHSILQRM RELTVQAAND TNVTVDRENL QKEVAELQSE INRISSQTQF NTKSLINGAV SKDADAMTFQ IGANSGQTIK LTIGSMDAKT LNVDKIKLDD TAANVTKQIA TIDNAINSVS KERAKLGANQ NRLEHTIANL DNSAENLQAA ESRIRDVDMA KEMMNFTKTN ILTQAAQAML AQANQAPQGV LQLLR | |
Greater than 90% as determined by SDS-PAGE. |