missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type F Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type F, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7209-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Botulinum neurotoxin type F Protein | |
Research Use Only | |
botF | |
Recombinant Protein | |
E. coli | |
MPVAINSFNY NDPVNDDTIL YMQIPYEEKS KKYYKAFEIM RNVWIIPERN TIGTNPSDFD PPASLKNGSS AYYDPNYLTT DAEKDRYLKT TIKLFKRINS NPAGKVLLQE ISYAKPYLGN DHTPIDEFSP VTRTTSVNIK LSTNVESSML LNLLVLGAGP DIFESCCYPV RKLIDPDVVY DPSNYGFGSI NIVTFSPEYE YTFNDISGGH NSSTESFIAD PAISLAHELI HALHGLYGAR GVTYEETIEV KQAPLMIAEK PIRLEEFLTF GGQDLNIITS AMKEKIYNNL LANYEKIATR LSEVNSAPPE YDINEYKDYF QWKYGLDKNA DGSYTVNENK FNEIYKKLYS FTESDLANKF KVKCRNTYFI KYEFLKVPNL LDDDIYTVSE GFNIGNLAVN NRGQSIKLNP KIIDSIPDKG LVEKIVKFCK SVIPRK | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |