missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type A Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7051-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003356619.1 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |
Botulinum neurotoxin type A Protein | |
Research Use Only | |
botA | |
Recombinant Protein | |
Yeast | |
MPFVNKQFNY KDPVNGVDIA YIKIPNVGQM QPVKAFKIHN KIWVIPERDT FTNPEEGDLN PPPEAKQVPV SYYDSTYLST DNEKDNYLKG VTKLFERIYS TDLGRMLLTS IVRGIPFWGG STIDTELKVI DTNCINVIQP DGSYRSEELN LVIIGPSADI IQFECKSFGH EVLNLTRNGY GSTQYIRFSP DFTFGFEESL EVDTNPLLGA GKFATDPAVT LAHELIHAGH RLYGIAINPN RVFKVNTNAY YEMSGLEVSF EELRTFGGHD AKFIDSLQEN EFRLYYYNKF KDIASTLNKA KSIVGTTASL QYMKNVFKEK YLLSEDTSGK FSVDKLKFDK LYKMLTEIYT EDNFVKFFKV LNRKTYLNFD KAVFKINIVP KVNYTIYDGF NLRNTNLAAN FNGQNTEINN MNFTKLKNFT GLFEFYKLLC VRGIIT | |
Greater than 90% as determined by SDS-PAGE. |