missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Bos taurus (Bovine) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6778-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045780

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001159540.1
TGF-beta 1 / TGFB1 Protein
Research Use Only
TGFB1
Recombinant Protein
E. coli
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS
Greater than 90% as determined by SDS-PAGE.
282089
10 μg
Not determined.
Partial (see sequence information for more details).
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers