missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Serpin A3-2 Protein
A DNA sequence encoding the Bos taurus (Bovine) Serpin A3-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9699-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139773.1 | |
Serpin A3-2 Protein | |
Research Use Only | |
SERPINA3-2 | |
Recombinant Protein | |
Yeast | |
LPENVVVKDQ HRRVDGHTLA SSNTDFAFSL YKQLALKNPN KNVILSPLSV SIALAFLSLG ARGSTLTEIL EGLKFNLTEI QEKEIHHSFQ HLLQALNQPS NQLQLSVGNA MFVQEELKLL DKFIEDAQVL YSSEAFPTNF RDSEAARSLI NDYVKNKTQG KIEELFKYLS PRTELVLVNY IYFKAQWKTP FDPKHTEQAE FHVSDNKTVE VPMMTLDLET PYFRDEELGC TLVELTYTSN DSALFILPDE GKMRDLEAKL TPETLTRWRN SLQPRRIHEL YLPKFSIKSN YELNDILSQL GIRKIFANAD LSGITGTADL VVSQVVHGAA LDVDEEGTEG VAATGIGIER TFLRIIVRVN RPFLIAVVLK DTQSIIFLGK VTNPSEA | |
Greater than 90% as determined by SDS-PAGE. |
100272170 | |
200 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |