missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Resistin / ADSF / RETN Protein
A DNA sequence encoding the Bos taurus (Bovine) Resistin / ADSF / RETN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6598-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_899206.1 | |
Resistin / ADSF / RETN Protein | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
QSLCPIDKAI SEKIQEVTTS LVPGAVRIIG LDCRSVTSRG SLVTCPSGFA VTGCTCGSAC GSWDVRAETT CHCQCAGMDW TGARCCRLHI Q | |
Greater than 90% as determined by SDS-PAGE. |
369020 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |