missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine PIGR Protein

A DNA sequence encoding the Bos taurus (Bovine) PIGR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6494-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023710

  • 2990.00 EUR / 1mg
Estimated Shipment: 27-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_776568.1
PIGR Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Bovine
His
Tris-based buffer, 50% glycerol
281401
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PIGR
Recombinant Protein
E. coli
ESRGLIKEQY EGRLALLTEP GNGTYTVILN QLTDQDTGFY WCVTDGDTRW ISTVELKVVQ GEPSLKVPKN VTAWLGEPLK LSCHFPCKFY SFEKYWCKWS NRGCSALPTQ NDGPSQAFVS CDQNSQVVSL NLDTVTKEDE GWYWCGVKEG PRYGETAAVY VAVESRVKGS QGAKQVKAAP AGAAIQSRAG EIQNKALLDP S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers