missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Interferon tau-1 Protein

A DNA sequence encoding the Bos taurus (Bovine) Interferon tau-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7076-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007300

  • 309.00 EUR / 10µg


Estimated Shipment
07-08-2020
Add to basket

Specifications

Specifications

NP_001015511.3
Interferon tau-1 Protein
10 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
317698
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFNT1
Recombinant Protein
E. coli
CYLSEDHMLG ARENLRLLAR MNRLSPHPCL QDRKDFGLPQ EMVEGNQLQK DQAISVLHEM LQQCFNLFYT EHSSAAWNTT LLEQLCTGLQ QQLEDLDACL GPVMGEKDSD MGRMGPILTV KKYFQGIHVY LKEKEYSDCA WEIIRVEMMR ALSSSTTLQK RLRKMGGDLN SL
Videos
Documents