missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Galectin-1 / LGALS1 Protein

A DNA sequence encoding the Bos taurus (Bovine) Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6293-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_786976.1
Galectin-1 / LGALS1 Protein
Research Use Only
LGALS1
Recombinant Protein
E. coli
ACGLVASNLN LKPGECLRVR GEVAADAKSF LLNLGKDDNN LCLHFNPRFN AHGDVNTIVC NSKDAGAWGA EQRESAFPFQ PGSVVEVCIS FNQTDLTIKL PDGYEFKFPN RLNLEAINYL SAGGDFKIKC VAFE
Greater than 90% as determined by SDS-PAGE.
326598
500 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers