missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine CCL20 / MIP-3 alpha Protein
A DNA sequence encoding the Bos taurus (Bovine) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.
352.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_776688.1 |
---|---|
Gene ID (Entrez) | 281666 |
Name | CCL20 / MIP-3 alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031521
|
enQuireBio™
QP7906-YE-100UG |
100 μg |
965.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071521
|
enQuireBio™
QP7906-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041521
|
enQuireBio™
QP7906-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061521
|
enQuireBio™
QP7906-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081521
|
enQuireBio™
QP7906-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051521
|
enQuireBio™
QP7906-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776688.1 | |
CCL20 / MIP-3 alpha Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281666 | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
Yeast | |
ASNFDCCLRY TERILHPSIL VGFTQQLANE ACDINAVVFY TRKKLAVCAD PKKKWVKQVV HMLSQRVKRM | |
Greater than 90% as determined by SDS-PAGE. |