missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Cathelicidin-4 Protein
A DNA sequence encoding the Bos taurus (Bovine) Cathelicidin-4, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_777252.1 |
---|---|
Gene ID (Entrez) | 282166 |
Name | Cathelicidin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097720
|
enQuireBio™
QP7211-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017730
|
enQuireBio™
QP7211-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007730
|
enQuireBio™
QP7211-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027730
|
enQuireBio™
QP7211-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037730
|
enQuireBio™
QP7211-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047730
|
enQuireBio™
QP7211-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_777252.1 | |
Cathelicidin-4 Protein | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
282166 | |
Research Use Only | |
CATHL4 | |
Recombinant Protein | |
E. coli | |
ILPWKWPWWP WRR | |
Greater than 90% as determined by SDS-PAGE. |