missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine ApoAI Protein

A DNA sequence encoding the Bos taurus (Bovine) ApoAI, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8986-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_776667.2
ApoAI Protein
Research Use Only
APOA1
Recombinant Protein
Yeast
DDPQSSWDRV KDFATVYVEA IKDSGRDYVA QFEASALGKQ LNLKLLDNWD TLASTLSKVR EQLGPVTQEF WDNLEKETAS LRQEMHKDLE EVKQKVQPYL DEFQKKWHEE VEIYRQKVAP LGEEFREGAR QKVQELQDKL SPLAQELRDR ARAHVETLRQ QLAPYSDDLR QRLTARLEAL KEGGGSLAEY HAKASEQLKA LGEKAKPVLE DLRQGLLPVL ESLKVSILAA IDEASKKLNA Q
Greater than 90% as determined by SDS-PAGE.
281631
200 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers