missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Agouti-signaling Protein

A DNA sequence encoding the Bos taurus (Bovine) Agouti-signaling protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7635-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069930

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_996674.1
Agouti-signaling Protein
Research Use Only
ASIP
Recombinant Protein
E. coli
HLAPEEKPRD ERNLKNNSSM NLLDFPSVSI VALNKKSKKI SRNEAEKKKR PSKRKAPMKN VARTRPPPPT PCVATRDSCK PPAPACCDPC AFCQCRFFRS ACSCRVLNPT C
Greater than 90% as determined by SDS-PAGE.
404192
50 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers