missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 1 subunit B Protein

A DNA sequence encoding the Enterobacteria phage H19B (Bacteriophage H19B) Shiga-like toxin 1 subunit B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6941-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006800

  • 1415.00 EUR / 200µg


Estimated Shipment
17-02-2020
Add to basket

Specifications

Specifications

Shiga-like toxin 1 subunit B Protein
200 μg
Not determined.
Full Length
Bacteriophage
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
stxB
Recombinant Protein
Yeast
TPDCVTGKVE YTKYNDDDTF TVKVGDKELF TNRWNLQSLL LSAQITGMTV TIKTNACHNG GGFSEVIFR
Videos
Documents