missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Recombination and repair Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Recombination and repair protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7355-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_049656.2 | |
Recombination and repair Protein | |
Research Use Only | |
UVSX | |
Recombinant Protein | |
E. coli | |
MSDLKSRLIK ASTSKLTAEL TASKFFNEKD VVRTKIPMMN IALSGEITGG MQSGLLILAG PSKSFKSNFG LTMVSSYMRQ YPDAVCLFYD SEFGITPAYL RSMGVDPERV IHTPVQSLEQ LRIDMVNQLD AIERGEKVVV FIDSLGNLAS KKETEDALNE KVVSDMTRAK TMKSLFRIVT PYFSTKNIPC IAINHTYETQ EMFSKTVMGG GTGPMYSADT VFIIGKRQIK DGSDLQGYQF VLNVEKSRTV KEKSKFFIDV KFDGGIDPYS GLLDMALELG FVVKPKNGWY AREFLDEETG EMIREEKSWR AKDTNCTTFW GPLFKHQPFR DAIKRAYQLG AIDSNEIVEA EVDELINSKV EKFKSPESKS KSAADLETDL EQLSDMEEFN E | |
Greater than 86.2% as determined by SDS-PAGE. |
1258704 | |
500 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |